Learn More
Sino Biological Anti-TROP-2 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10428MM01100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human TROP2 extracellular domain (rh TROP2; Catalog#10428-H08H; NP_002344.2; Met 1-Thr 274). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-5688