Learn More
Sino Biological Recombinant Anti-TROP-2 Antibody (FITC), Rabbit Monoclonal
Supplier:  Sino Biological 10428R001F25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human TROP2 / TACSTD2 (rh TROP2 / TACSTD2; Catalog#10428-H08H; NP_002344.2; Met1-Thr274) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-172-5704