Learn More
Sino Biological Anti-TROP-2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10428RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human TROP2 extracellular domain (rh TROP2; Catalog#10428-H08H; NP_002344.2; Met 1-Thr 274). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-172-5709