Learn More
Sino Biological Anti-TROP-2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10428T24100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human TROP-2 / TACSTD2 (rh TROP-2 / TACSTD2; Catalog#10428-H08H; NP_002344.2; Met1-Thr274). TROP-2 / TACSTD2 specific IgG was purified by Human TROP-2 / TACSTD2 affinity chromatography.
Catalog No. 50-172-5715