Learn More
Sino Biological Recombinant Human IGFBP-3 Protein (His Tag)
Supplier:  Sino Biological 10430H07H1
Encompass_Preferred
A DNA sequence encoding the mature form of human IGFBP3 (NP_000589.2) (Gly28-Lys291) was fused with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-4928