Learn More
Sino Biological Recombinant Anti-IGFBP-3 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10430R010100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human IGFBP3 / IGFBP-3 (rh IGFBP3 / IGFBP-3; Catalog#10430-H07H; NP_000589.2; Gly28-Lys291).
Catalog No. 50-172-5725