Learn More
Sino Biological Anti-IGFBP-3 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10430RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human IGFBP3 (rh IGFBP3; Catalog#10430-H07H; NP_000589.2; Gly 28-Lys 291). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-172-5727