Learn More
Sino Biological Anti-IGFBP-3 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10430T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human IGFBP3 / IGFBP-3 (rh IGFBP3 / IGFBP-3; Catalog#10430-H07H; NP_000589.2; Gly28-Lys291). IGFBP3 / IGFBP-3 specific IgG was purified by Human IGFBP3 / IGFBP-3 affinity chromatography.
Catalog No. 50-172-5733