Learn More
Sino Biological Recombinant Human TWEAKR Protein (hFc Tag)
Supplier:  Sino Biological 10431H01H100
Encompass_Preferred
A DNA sequence encoding the human TNFRSF12A isoform 1 (Q9NP84-1) extracellular domain (Glu 28-Trp 79) was fused with the Fc region of human IgG1 at the N-terminus.
Catalog No. 50-161-4933