Learn More
Sino Biological Anti-VE-Cadherin Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10433MM02100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human VE-Cadherin / CD144 (CAA56306.1; Met1-Gln593). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-5748