Learn More
Sino Biological Recombinant Anti-VE-Cadherin Antibody (FITC), Rabbit Monoclonal
Supplier:  Sino Biological 10433R048F100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human VE-Cadherin / CD144 / CDH5 (rh VE-Cadherin / CD144 / CDH5; CAA56306.1; Met1-Gln593) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-172-5762