Learn More
Sino Biological Anti-VE-Cadherin Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10433RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human VE-Cadherin / CD144 / CDH5 (rh VE-Cadherin / CD144 / CDH5; CAA56306.1; 1M-593Q). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-172-5766