Learn More
Sino Biological Anti-VE-Cadherin Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10433T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human VE-Cadherin / CD144 / CDH5 (rh VE-Cadherin / CD144 / CDH5; CAA56306.1; Met1-Gln593). VE-Cadherin / CD144 / CDH5 specific IgG was purified by Human VE-Cadherin / CD144 / CDH5 affinity chromatography.
Catalog No. 50-172-5769