Learn More
Sino Biological Recombinant Human Cystatin B Protein (His Tag)
Supplier:  Sino Biological 10435H07E100
Encompass_Preferred
A DNA sequence encoding the human CSTB (P04080) (Met 2-Phe 98) was expressed, with a polyhistide tag at the N-terminus.
Catalog No. 50-161-4937