Learn More
Sino Biological Anti-Cystatin B Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10435RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Cystatin B / CSTB (rh CSTB; Catalog#10435-H07E; P04080; Met2-Phe98). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-172-5778