Learn More
Sino Biological Recombinant Human Cyclophilin A Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10436H08E100
Encompass_Preferred
A DNA sequence encoding the human PPIA (P62937) (Met 1-Glu 165) was expressed, with a polyhistide tag at the C-terminus.
Catalog No. 50-161-4939