Learn More
Sino Biological Anti-Cyclophilin A Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10436T52100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Cyclophilin A (rh Cyclophilin A; Catalog#10436-H08E; P62937; Met1-Glu165). Cyclophilin A specific IgG was purified by Human Cyclophilin A affinity chromatography.
Catalog No. 50-172-5784