Learn More
Sino Biological Anti-NKX1-2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 104372T36100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Mouse NKX1-2, and purified by antigen affinity chromatography.
Catalog No. 50-172-5787