Learn More
Sino Biological Recombinant Human CST6 Protein (His Tag)
Supplier:  Sino Biological 10438H08H20
Encompass_Preferred
A DNA sequence encoding the human cystatin E/M protein (NP_001314.1) (Met 1-Met 149) was expressed with a C-terminal polyhistidine-tag.
Catalog No. 50-161-4944