Learn More
Sino Biological Recombinant Anti-CST6 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10438R034100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human Cystatin E / CST6 (rh Cystatin E / CST6; Catalog#10438-H08H; NP_001314.1; Met 1-Met 149).
Catalog No. 50-172-5812