Learn More
Sino Biological Recombinant Human FLT3 Protein (T227M, ECD, His Tag), HPLC-verified
Supplier:  Sino Biological 10445H08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of human FLT3 (P36888-1, with natural variant Thr 227 Met) (Met1-Asn541) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-4955