Learn More
Sino Biological Anti-FLT3 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10445RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human FLT3 (rh FLT3; Catalog#10390-H08H;AAL65157.1; Met 1-Arg 200). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-172-5857