Learn More
Sino Biological Recombinant Human BMPR1A/ALK-3 Protein (His Tag)
Supplier:  Sino Biological 10446H08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Arg 152) of human ALK3 (NP_004320.2) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-4959