Learn More
Sino Biological Anti-BMPR1A/ALK-3 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10446T24100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human ALK-3 / BMPR1A (rh ALK-3 / BMPR1A; Catalog#10446-H08H; NP_004320.2; Met1-Arg152). ALK-3 / BMPR1A specific IgG was purified by Human ALK-3 / BMPR1A affinity chromatography.
Catalog No. 50-172-5863