Learn More
Sino Biological Recombinant Human Annexin V/ANXA5 Protein, HPLC-verified
Supplier:  Sino Biological 10448HNAE100
Encompass_Preferred
A DNA sequence encoding the human ANXA5 (P08758) (Met 1-Asp 320) was expressed and purified.
Catalog No. 50-161-4963