Learn More
Sino Biological Anti-Annexin V/ANXA5 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10448T52100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human ANXA5 (rh ANXA5; Catalog#10448-HNAE; P08758; Met1-Asp320). ANXA5 specific IgG was purified by Human ANXA5 affinity chromatography.
Catalog No. 50-172-5875