Learn More
Sino Biological Anti-E2f6 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 104488T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human E2f6, and purified by antigen affinity chromatography.
Catalog No. 50-172-5866