Learn More
Sino Biological Recombinant Human ARSA Protein (His Tag)
Supplier:  Sino Biological 10449H08H50
Encompass_Preferred
A DNA sequence encoding the human Arylsulfatase A (P15289-1) (Met1-Ala507) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-4968