Learn More
Sino Biological Anti-ARSA Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10449RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human ARSA (rh ARSA; Catalog#10449-H08H; P15289-1; Met 1-Ala 507). ARSA specific IgG was purified by human ARSA affinity chromatography.
Catalog No. 50-172-5888