Learn More
Sino Biological Anti-Oncostatin M/OSM Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10452MM03100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Oncostatin M / OSM (rh Oncostatin M / OSM; Catalog#10452-H08H; P13725; Met1-Arg221). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-5912