Learn More
Sino Biological Anti-Oncostatin M/OSM Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10452RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Oncostatin M / OSM (rh Oncostatin M / OSM; Catalog#10452-H08H; P13725; Met 1-Arg 221). Oncostatin M / OSM specific IgG was purified by Human Oncostatin M / OSM affinity chromatography.
Catalog No. 50-172-5920