Learn More
Sino Biological Anti-Somatostatin Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 104537T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human SST, and purified by antigen affinity chromatography.
Catalog No. 50-172-5923