Learn More
Sino Biological Anti-BCL-xL/BCL2L1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10455T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human BCL2L1 / Bcl-XL (rh BCL2L1 / Bcl-XL; Catalog#10455-H08E; NP_612815.1; Met1-Arg212). BCL2L1 / Bcl-XL specific IgG was purified by Human BCL2L1 / Bcl-XL affinity chromatography.
Catalog No. 50-172-5934