Learn More
Sino Biological Recombinant Human TGF-beta RI Protein (aa 200-503, His & GST Tag)
Supplier:  Sino Biological 10459H20B50
Encompass_Preferred
A DNA sequence encoding the human ALK5 (P36897-1) (Thr200-Mey503) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.
Catalog No. 50-161-4990