Learn More
Sino Biological Recombinant Human BMPRIB Protein (aa 149-502, His & GST Tag), HPLC-verified
Supplier:  Sino Biological 10460H20B50
Encompass_Preferred
A DNA sequence encoding the human ALK6 (NP_001194.1) cytoplasmic domain (Arg 149-Leu 502) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.
Catalog No. 50-161-4994