Learn More
Sino Biological Recombinant Human FAP Protein (ECD, His Tag), HPLC-verified
Supplier:  Sino Biological 10464H07H50
Encompass_Preferred
A DNA sequence encoding the human FAP (Q12884-1) extracellular domain (Leu26-Asp760) was fused with the polyhistine tag at the N-terminus.
Catalog No. 50-161-5003