Learn More
eMolecules​ 10465-78-8 | 1,1'-Azobis(N,N-dimethylformamide) | Oakwood Chemicals | MFCD00008318 | 172.188 | C6H12N4O2 | 95.000 | CN(C)C(=O)\N=N\C(=O)N(C)C | 100g | 480133273
Supplier:  eMolecules​ 480133273
Encompass_Preferred
1,1'-Azobis(N,N-dimethylformamide) | Oakwood Chemicals | 10465-78-8 | MFCD00008318 | 172.188 | C6H12N4O2 | 95.000 | CN(C)C(=O)\N=N\C(=O)N(C)C | 100g | 480133273
Catalog No. 50-004-89146