Learn More
Sino Biological Recombinant Human Nogo Receptor/RTN4R Protein (His & hFc Tag)
Supplier:  Sino Biological 10466H03H100
Encompass_Preferred
A DNA sequence encoding the mature form of human RTN4R (NP_075380.1) (Met 1-Ser 447) was fused with the C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-5009