Learn More
Sino Biological Recombinant Human Nogo Receptor/RTN4R Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10466H08H100
Encompass_Preferred
A DNA sequence encoding the human RTN4R (Q9BZR6) (Met1-Ser447) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-5011