Learn More
Sino Biological Recombinant Human MMP-3 Protein, HPLC-verified
Supplier:  Sino Biological 10467HNAE100
Encompass_Preferred
A DNA sequence encoding the human MMP3 (AAA36321.1) N-terminal fragment (Tyr 18-Thr 272) was expressed and purified.
Catalog No. 50-161-5013