Learn More
Sino Biological Recombinant Human BID Protein
Supplier:  Sino Biological 10468HNCE100
Encompass_Preferred
A DNA sequence encoding the human BID isoform 1 (P55957-1) (Met 1-Asp 195) was expressed and purified, with additional two amino acids (Gly & Pro) at the N-terminus.
Catalog No. 50-161-5016