Learn More
eMolecules​ 1046816-75-4 | (2-CHLOROPYRIMIDIN-5-YL)METHANOL | AstaTech | MFCD16659172 | 144.560 | C5H5ClN2O | 95.000 | OCc1cnc(Cl)nc1 | 1g | 112533451
Catalog No. 5000492756 Shop All eMolecules Inc. Products
Encompass_Preferred
(2-CHLOROPYRIMIDIN-5-YL)METHANOL | AstaTech | 1046816-75-4 | MFCD16659172 | 144.560 | C5H5ClN2O | 95.000 | OCc1cnc(Cl)nc1 | 1g | 112533451
Catalog No. 50-004-92756 Supplier eMolecules​ Supplier No. 112533451
(2-CHLOROPYRIMIDIN-5-YL)METHANOL | AstaTech | 1046816-75-4 | MFCD16659172 | 144.560 | C5H5ClN2O | 95.000 | OCc1cnc(Cl)nc1 | 1g | 112533451