Learn More
Sino Biological Recombinant Human Frizzled-5 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10473H08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Pro 167) of human frizzled 5 (NP_003459.2) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-5026