Learn More
Sino Biological Recombinant Human Eotaxin/CCL11 Protein (His Tag)
Catalog No. 501615029
Encompass_Preferred
A DNA sequence encoding the human CCL11 (P51671) (Gly 24-Pro 97) was expressed, with a polyhistide tag at the N-terminus.
Catalog No. 50-161-5029 Supplier Sino Biological Supplier No. 10474H07E50
Add to Cart
Add to Cart
A DNA sequence encoding the human CCL11 (P51671) (Gly 24-Pro 97) was expressed, with a polyhistide tag at the N-terminus.