Learn More
Sino Biological Recombinant Human CCL21/6Ckine Protein
Supplier:  Sino Biological 10477HNAB50
Encompass_Preferred
A DNA sequence encoding the human CCL21 (O00585) (Met 1-Pro 134) was expressed and purified.
Catalog No. 50-161-5038