Learn More
Sino Biological Recombinant Human Cystatin A Protein (His Tag)
Supplier:  Sino Biological 10479H07E100
Encompass_Preferred
A DNA sequence encoding the human CSTA (P01040) (Ile 2-Phe 98) was expressed with a polyhistide tag at the N-terminus.
Catalog No. 50-161-5041