Learn More
Sino Biological Recombinant Human TCCR/IL-27R alpha Protein (hFc Tag), HPLC-verified
Supplier:  Sino Biological 10489H02H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Arg 514) of human IL27Rα (NP_004834.1) was fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-5064