Learn More
Sino Biological Recombinant Human LEC/CCL16 Protein (His Tag)
Supplier:  Sino Biological 10491H08Y50
Encompass_Preferred
A DNA sequence encoding human CCL16 (NP_004581.1) (Gln24-Gln120) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-5073