Learn More
Med Vet International 104mm Titanium 8-Hole Forkless TTA Plate, J1072TF8
Catalog No. 5000583279
Encompass_Preferred
Standard TTA plates rely on a barbed for system for attachment onto the tibial crest. Many surgeons consider that the hardest part of the TTA procedure is preparing for and placing the fork. Forkless TTA plates are screwed onto the tibial crest using 2.4 screws. The distal part of the forkless plate is secured using 2.7 or 3.5mm screws as appropriate. The rest of the pro- cedure is as per the standard technique. Forkless TTA Plate. 104mm
Catalog No. 50-005-83279 Supplier Med Vet International Supplier No. JOR1072TF8
Standard TTA plates rely on a barbed for system for attachment onto the tibial crest. Many surgeons consider that the hardest part of the TTA procedure is preparing for and placing the fork. Forkless TTA plates are screwed onto the tibial crest using 2.4 screws. The distal part of the forkless plate is secured using 2.7 or 3.5mm screws as appropriate. The rest of the pro- cedure is as per the standard technique. Forkless TTA Plate. 104mm