Learn More
Inorganic Ventures 10 CONDUCTIVITY 500mL
SDP
Catalog No. NC0874059
Encompass_Preferred
Inorganic Ventures' 10 mhos/cm Conductivity at 25C is a 500mL certified reference material set in a water matrix for stability. This standard is Traceable to NIST and manufactured in accordance with our stringent Quality Assurance guidelines. See Certificate of Analysis for expiration. Custom conductivity standards are available upon request.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. NC0874059 Supplier Inorganic Ventures Supplier No. CON1025500ML
Inorganic Ventures' 10 mhos/cm Conductivity at 25C is a 500mL certified reference material set in a water matrix for stability. This standard is Traceable to NIST and manufactured in accordance with our stringent Quality Assurance guidelines. See Certificate of Analysis for expiration. Custom conductivity standards are available upon request.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search